Få mere at vide
myo-Inositol Trispyrophosphate Hexasodium Salt, TRC
Artikelnummer. 30010086
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010086 10 mg
30008245 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010086 Leverandør Toronto Research Chemicals Leverandørnr. TRCI66602010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23103-35-7
Molekylær formel C6 H6 O21 P6 . 6[Na]
Molekylvægt (g/mol) 737.88
Synonym ITTP Hexasodium Salt, myo-Inositol Cyclic 1,2:3,4:5,6-Tris(dihydrogen pyrophosphate)) Hexasodium Salt, myo-Inositol Cyclic 1,2:3,4:5,6-Tris(P,P'-dihydrogen diphosphate) Hexasodium Salt
SMIL O=P1(O[C@]2([H])[C@@]3([H])[C@@](OP(OP(O3)(O[Na])=O)(O[Na])=O)([H])[C@]4([H])[C@@](OP(OP(O4)(O[Na])=O)(O[Na])=O)([H])[C@@]2([H])OP(O1)(O[Na])=O)O[Na]
Tekniske data
Kemisk navn eller materiale myo-Inositol Trispyrophosphate Hexasodium Salt
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C6H12O21P6.6Na/c7-28(8)19-1-2(20-29(9,10)25-28)4-6(24-33(17,18)27-32(15,16)23-4)5-3(1)21-30(11,12)26-31(13,14)22-5;;;;;;/h1-6H,(H,7,8)(H,9,10)(H,11,12)(H,13,14)(H,15,16)(H,17,18);;;;;;/q;6*+1/p-6/t1-,2-,3-,4+,5-,6-;;;;;;/m0....../s1
Formel vægt 737.721
Procent renhed >95%