Få mere at vide
myo-Inositol-d6, TRC
Artikelnummer. 30408355
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30408355 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408355 Leverandør Toronto Research Chemicals Leverandørnr. TRCI66599710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale myo-Inositol-1,2,3,4,5,6-d6
CAS 68922-44-1
Molekylær formel C6 2H6 H6 O6
Mængde 10 mg
Synonym myo-[1,2,3,4,5,6-2H2]Inositol, myo-Inositol-1,2,3,4,5,6-d6, myo-Inositol-C-d6, myo-Inositol-1,2,3,4,5,6-C-d6 (9CI), Inositol-d6 (6CI)
InChI formel InChI=1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4+,5-,6-/i1D,2D,3D,4D,5D,6D
SMIL [2H][C@@]1(O)[C@@]([2H])(O)[C@@]([2H])(O)[C@@]([2H])(O)[C@]([2H])(O)[C@@]1([2H])O
IUPAC navn 1,2,3,4,5,6-hexadeuteriocyclohexane-1,2,3,4,5,6-hexol
Molekylvægt (g/mol) 186.19
Formel vægt 186.101