missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin Protein Zero Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-13353
Additional Details : Weight : 0.00970kg
Description
Myelin Protein Zero Polyclonal antibody specifically detects Myelin Protein Zero in Mouse, Rat samples. It is validated for Western BlotSpecifications
Myelin Protein Zero | |
Polyclonal | |
PBS, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
Charcot-Marie-Tooth neuropathy 1B, CHM, CMT1, CMT1B, CMT2I, CMT2J, CMT4E, CMTDI3, HMSNIB, MPP, Myelin peripheral protein, myelin protein P0, myelin protein zeroDSS, P0 | |
Recombinant protein encompassing a sequence within the center region of mouse Myelin Protein Zero. The exact sequence is proprietary. | |
100 μg | |
Neuronal Cell Markers, Oligodendrocyte Cell Markers | |
4359 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |