Få mere at vide
Mycophenolate Mofetil-d4, TRC
Artikelnummer. 30040945
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30040945 50 mg
30040736 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30040945 Leverandør Toronto Research Chemicals Leverandørnr. TRCM83145250MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1132748-21-0
Molekylær formel C23 2H4 H27 N O7
Molekylvægt (g/mol) 437.52
Synonym Mycophenolate mofetil-D4 (morpholinylethyl-1,1,2,2-D4), 4-Hexenoic acid, 6-(1,3-dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-, 2-(4-morpholinyl)ethyl-1,1,2,2-d4 ester, (4E)-, Mycophenolate mofetil-d4, (4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid 2-(4-morpholinyl)ethyl-1,1,2,2-d4 ester
IUPAC navn (1,1,2,2-tetradeuterio-2-morpholin-4-ylethyl) (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
SMIL [2H]C([2H])(OC(=O)CC\C(=C\Cc1c(O)c2C(=O)OCc2c(C)c1OC)\C)C([2H])([2H])N3CCOCC3
Tekniske data
Kemisk navn eller materiale Mycophenolate mofetil-D4
Mængde 50 mg
InChI formel InChI=1S/C23H31NO7/c1-15(5-7-19(25)30-13-10-24-8-11-29-12-9-24)4-6-17-21(26)20-18(14-31-23(20)27)16(2)22(17)28-3/h4,26H,5-14H2,1-3H3/b15-4+/i10D2,13D2
Formel vægt 437.2352