Få mere at vide
Honeywell™ Mutex E Light Protective Clothing
Artikelnummer. 15683346
Størrelse:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Pakningsstørrelse:
Hver
Artikelnummer. 15683346
Brand: Honeywell™ 4500101S
Breathable, single-use protective clothing designed to protect against particle contamination and chemical splashes.
  • Antistatic
  • Silicone and latex free
  • Excellent ergonomics and fitting
    • 3-piece hood
    • Special crotch design
    • Outside flap and dual slider sipper
    • Breathability
    • 3-layer polypropylene
TRUSTED_SUSTAINABILITY
Tekniske data
Produkttype Chemical-resistant Coveralls
Størrelse Small
Køn Unisex
Materiale 3-layer Polypropylene
Farve White
Beskyttelsesklasse Protection type: 5, 6
Tykkelse 56 gsm