Få mere at vide
Mutaprodenafil, TRC
Artikelnummer. 30481863
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30481863 25 mg -
30468312 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30481863 Leverandør Toronto Research Chemicals Leverandørnr. TRCM82000025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1387577-30-1
Molekylær formel C27H35N9O5S2
Molekylvægt (g/mol) 629.75
Synonym rel-5-[5-[[(3R,5S)-3,5-dimethyl-1-piperazinyl]sulfonyl]-2-ethoxyphenyl]-1-methyl-7-[(1-methyl-4-nitro-1H-imidazol-5-yl)thio]-3-propyl-1H-pyrazolo[4,3-d]pyrimidine
SMIL O=S(C1=CC=C(OCC)C(C2=NC3=C(N(C)N=C3CCC)C(SC4=C([N+]([O-])=O)N=CN4C)=N2)=C1)(N5C[C@H](C)N[C@H](C)C5)=O
Tekniske data
Kemisk navn eller materiale Mutaprodenafil
Mængde 25 mg
InChI formel InChI=1S/C27H35N9O5S2/c1-7-9-20-22-23(34(6)32-20)26(42-27-25(36(37)38)28-15-33(27)5)31-24(30-22)19-12-18(10-11-21(19)41-8-2)43(39,40)35-13-16(3)29-17(4)14-35/h10-12,15-17,29H,7-9,13-14H2,1-6H3/t16-,17+