missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MUSK Recombinant Human Protein

MUSK plays an important and selective role in many aspects of skeletal muscle development and function.

Brand:  Invitrogen™ PV4113

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10775443

  • 57500.00 DKK / 1mg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MUSK plays an important role in skeletal muscle development through the initial step of postsynaptic differentiation and formation of end-plate bands in cooperation with Rapsyn.

Specifications

Specifications

NP_005583.1
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MUSK
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
69.4kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
MUSK Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers