missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Musashi-2 Antibody (960121) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3255JF669
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Tekniske data
| Musashi-2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2 | |
| E. coli-derived recombinant human Musashi-2, Met1-His328, Accession # Q96DH6 | |
| 0.1 mL | |
| Neuronal Cell Markers | |
| 124540 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 960121 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion