Få mere at vide
Mupirocin Descarboxylic Acid (>90%), TRC
Artikelnummer. 30366534
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366534 5 mg
30364367 50 mg
30362697 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366534 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2269855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 153900-96-0
Molekylær formel C27H48O7
Molekylvægt (g/mol) 484.67
Synonym Mucirocin Impurity 1, Decyl [2E,8[2S,3S(1S,2S)]]-5,9-Anhydro-2,3,4,8-tetradeoxy-8-[[3-(2-hydroxy-1-methylpropyl)oxiranyl]methyl]-3-methyl-L-talo-Non-2-enonic Acid Ester
IUPAC navn decyl (E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoate
SMIL CCCCCCCCCCOC(=O)\C=C(/C)\C[C@@H]1OC[C@H](C[C@@H]2O[C@H]2[C@@H](C)[C@H](C)O)[C@@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale Mupirocin Descarboxylic Acid
Mængde 5 mg
InChI formel InChI=1S/C27H48O7/c1-5-6-7-8-9-10-11-12-13-32-24(29)15-18(2)14-22-26(31)25(30)21(17-33-22)16-23-27(34-23)19(3)20(4)28/h15,19-23,25-28,30-31H,5-14,16-17H2,1-4H3/b18-15+/t19-,20-,21-,22-,23-,25+,26-,27-/m0/s1
Formel vægt 484.34