Få mere at vide
Witeg™ Magnetic Stirrer Hotplate SMHS 6-Place
Topplademateriale:
Keramisk belagt aluminium
Beskrivelse
- Each place individually adjustable
- Digital Jog-Dial controller for each place
- Heat up to 350°C
- Up to 1500 rpm
- Ceramic-coated aluminum plate with 6 places 140 x 140 mm
- Heat-, chemical- and restricted acid-resistant
- No liquid can get into the body
- Powder-coated steel body
- Easy to clean
- 30 mm stirrer bar included (6x)
- Up to 3 temperature probes possible
- CE certified and unique serial number for tracing
- Timer 99 hr. 59 min. with timer-end signal
- Error status
- Locking mode
- Operation LEDs
- Jog-Dial controller with touch-button
Tekniske data
Tekniske data
| Topplademateriale | Keramisk belagt aluminium |
| Spænding | 230 V |
| Inkluderer | 6 Stir Bars |
| Kropsmateriale | Powder-coated steel |
| Controller type | Digital |
| Størrelse | 5.5 x 5.5 in. |
| Temperatur (metrisk) | Ambient to 350°C |
| Watt | 6 x 400 W |
| Type | Stirring Hotplate |
| Form | Square |
| Vis mere |