Learn More
Multi-Element Organometallic Oil Standards 23 Element, 500μg/g; SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
2915.00 DKK - 4510.00 DKK
Specifications
Chemical Name or Material | 23 Element Oil Standard |
---|---|
Packaging | Triple-leached LDPE bottle |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
Analyte or Component Names | Al,Ba,B |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440359
|
SPEX Certiprep
S23-500Z |
50 g |
2915.00 DKK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15236386
|
SPEX Certiprep
S23-500Y |
100 g |
4510.00 DKK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 500ÎĽg/g |
Al | 500ÎĽg/g |
B | 500ÎĽg/g |
Ba | 500ÎĽg/g |
Ca | 500ÎĽg/g |
Cd | 500ÎĽg/g |
Cr | 500ÎĽg/g |
Cu | 500ÎĽg/g |
Fe | 500ÎĽg/g |
K | 500ÎĽg/g |
Mg | 500ÎĽg/g |
Mn | 500ÎĽg/g |
Mo | 500ÎĽg/g |
Na | 500ÎĽg/g |
Ni | 500ÎĽg/g |
P | 500ÎĽg/g |
Pb | 500ÎĽg/g |
Sb | 500ÎĽg/g |
Si | 500ÎĽg/g |
Sn | 500ÎĽg/g |
Ti | 500ÎĽg/g |
V | 500ÎĽg/g |
Zn | 500ÎĽg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
23 Element Oil Standard | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Al, Ba, B | |
Petrochemical Testing |
Triple-leached LDPE bottle | |
Al,Ba,B | |
Certified Reference Material | |
Base Oil 75 |
Safety and Handling
May cause an allergic skin reaction
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only