Få mere at vide
Multi-Cation Standard 1: Li+ @ 50; Na+ @ 200; Mg+2, NH4+ @ 250; Ca+2, K+ @ 500 μg/mL in dil. HNO3, VHG™
Artikelnummer. 30003159
Skift visning
Mængde:
100 ml
250 ml
500 ml
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003159 100 ml
30003125 500 ml
30003151 250 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Artikelnummer. 30003159 Leverandør VHG Labs Leverandørnr. VHGICM5A100
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Multi-Cation Standard 1: Li+ @ 50; Na+ @ 200; Mg+2, NH4+ @ 250; Ca+2, K+ @ 500 μg/mL in dil. HNO3
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lithium Ion/Sodium Ion/Ammonium/Magnesium Ion 2+/Calcium Ion 2+/Potassium Ion
CAS 14798-03-9, 14127-61-8
Metoder Ion Chromatography
Analyt- eller komponentnavne Lithium Ion/Sodium Ion/Ammonium/Magnesium Ion 2+/Calcium Ion 2+/Potassium Ion
Forsendelsestilstand Room Temperature
Certificeringer/compliance ISO 17034
Opløsningsmiddel eller Matrix Dilute HNO3
Mængde 100 ml
Koncentration eller sammensætning (efter analyt eller komponenter) Lithium Ion 50 μg/mL/Sodium Ion 200 μg/mL/Ammonium 250 μg/mL/Magnesium Ion 2+ 250 μg/mL/Calcium Ion 2+ 500 μg/mL/Potassium Ion 500 μg/mL