Få mere at vide
Multi-Anion Standard 4: F- @ 100; Cl- @ 200; Br-, NO3-, SO4(-2) @ 400; PO4(-3) @ 600 μg/mL in H2O, VHG™
Artikelnummer. 17938104
Skift visning
Mængde:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17938104 500 ml
17969734 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17938104 Leverandør VHG Labs Leverandørnr. VHGICM4500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Multi-Anion Standard 4: F- @ 100; Cl- @ 200; Br-, NO3-, SO4(-2) @ 400; PO4(-3) @ 600 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fluoride/Chloride/Bromide/Nitrate/Sulfate/Phosphate
CAS 16984-48-8, 16887-00-6, 24959-67-9, 14797-55-8, 14808-79-8, 14265-44-2
Metoder Ion Chromatography
Analyt- eller komponentnavne Fluoride/Chloride/Bromide/Nitrate/Sulfate/Phosphate
Forsendelsestilstand Room Temperature
Certificeringer/compliance ISO 17034
Opløsningsmiddel eller Matrix Water
Mængde 500 ml
Koncentration eller sammensætning (efter analyt eller komponenter) Fluoride 20 μg/mL /Chloride 30 μg/mL /Bromide 400 μg/mL /Nitrate 400 μg/mL /Sulfate 400 μg/mL /Phosphate 600 μg/mL