Få mere at vide
Multi-Anion Standard 2: F-, Cl-, SO4(-2) @ 100 μg/mL in H2O, VHG™
Artikelnummer. 17959734
Skift visning
Mængde:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17959734 100 ml
17929634 500 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17959734 Leverandør VHG Labs Leverandørnr. VHGICM2100
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Multi-Anion Standard 2: F-, Cl-, SO4(-2) @ 100 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Chloride/Fluoride/Sulfate
CAS 16887-00-6, 16984-48-8, 14808-79-8
Metoder Ion Chromatography
Analyt- eller komponentnavne Chloride/Fluoride/Sulfate
Forsendelsestilstand Room Temperature
Certificeringer/compliance ISO 17034
Opløsningsmiddel eller Matrix Water
Mængde 100 ml
Koncentration eller sammensætning (efter analyt eller komponenter) Chloride 100 μg/mL/Fluoride 100 μg/mL/Sulfate 100 μg/mL