Learn More
Multi 4 elements solution for ICP 0.01mg/L, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical MS 002 500
Additional Details : Weight : 0.50000kg
Specifications
Four Components | |
Bottle | |
Limit Detection Multi-component Verification Solution | |
1% HNO3 |
0.01mg/L | |
500 mL | |
Be, Co, In, U | |
Liquid |