Learn More
Nabertherm™ Muffle Furnaces with Lift Door
For research and laboratory applications
Brand:  Nabertherm™ L-152H1RN1
Additional Details : Weight : 60.00000kg
 View more versions of this product
Product Code. 15486702
  • 27221.54 DKK / Each
Estimated Shipment
07-10-2020
Add to basket
Description
Description
  • Insulation made of non-classified fiber material
  • Housing made of sheets of textured stainless steel
  • Textured stainless steel sheet housing
  • Equipped with lift door (LT)
  • Dual shell housing for low external temperatures and high stability
  • Adjustable air inlet integrated into the door, exhaust air outlet in the rear wall of furnace
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low-noise operation
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
  • Supplied with B410 controller with four programs and five freely programmable ramps, or C450 with 10 freely adjustable programs and 20 segments
  • Both controllers have integrated USB interface and offer plain text display in different languages
  • Available in different sizes and temperature ranges
Additional equipment
  • Chimney, chimney with fan or catalytic converter
  • Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load
  • Protective gas connection to purge with non-flammable protective or reaction gases
  • Manual or automatic gas supply system
Specifications
Specifications
C450
1-Phase
3.5 kw
9 L
240 x 230 x 170 mm
515 + 240 mm (Including opened lift door)
415 mm
Muffle Furnace with Lift Door