Få mere at vide
Nabertherm™ Muffle Furnace LT5/13/C550 Tmax1300 with Lift Door
Shop alle Nabertherm produkter
Temperatur (metrisk):
1300 °C
Beskrivelse
- Heating elements on support tubes radiating freely into the furnace chamber provide for particularly short heating times for these muffle furnaces. Thanks to their robust lightweight refractory brick insulation, they can reach a maximum working temperature of 1300°C. These muffle furnaces thus represent an interesting alternative to the familiar L(T) ../12 models, when you need a higher application temperature.
- Standard Equipment
- Tmax 1300°C
- Heating from two sides
- Heating elements on support tubes ensure free heat radiation and a long service life
- Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
- Dual shell ventilated housing made of textured stainless steel sheets for low surface temperature and high stability
- Optional flap door (L) which can be used as work platform or lift door (LT) with hot surface facing away from the operator
- Adjustable air inlet in the furnace door
- Exhaust air outlet in rear wall of furnace
- Controller with touch operation B510 (5 programs with each 4 segments)
- Solid state relays provide for lownoise operation
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Tekniske data
Tekniske data
| Temperatur (metrisk) | 1300 °C |
| Type | Muffle Furnace with Lift Door |
| Spænding | 220/240 V |