Learn More
Mucin, MP Biomedicals™ Product Code.: 11444931
Mucin, MP Biomedicals™
Product Code. 11444931 Shop All MP Biomedical Products
100 mg, 100mg
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 11444931
Brand: MP Biomedicals 0215574280
Chemical Identifiers
CAS 84195-52-8
Specifications
Color Tan
Quantity 100 mg
Formula Weight ≈400,0002
Physical Form Flaky Powder
Chemical Name or Material Mucin
Product Content Correction
Product Title
Mucin, MP Biomedicals™ >