missing translation for 'onlineSavingsMsg'
Learn More

Mucic Acid 98%, Thermo Scientific Chemicals

Catalog Number p-170156
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
526-99-8
C6H10O8
210.138
MFCD00004239
DSLZVSRJTYRBFB-DUHBMQHGSA-N
galactaric acid, mucic acid, 2r,3s,4r,5s-2,3,4,5-tetrahydroxyhexanedioic acid, saccharolactic acid, mucicacid, unii-e149j5otif, d-galactaric acid, galactosaccharic acid, galactarate, meso-galactaric acid
3037582
CHEBI:30852
(2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
526-99-8
C6H10O8
210.138
MFCD00004239
DSLZVSRJTYRBFB-DUHBMQHGSA-N
galactaric acid, mucic acid, 2r,3s,4r,5s-2,3,4,5-tetrahydroxyhexanedioic acid, saccharolactic acid, mucicacid, unii-e149j5otif, d-galactaric acid, galactosaccharic acid, galactarate, meso-galactaric acid
3037582
CHEBI:30852
(2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 03, IV, 1292
Merck Index 15, 4361
Formula Weight 210.14
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Mucic acid, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'