missing translation for 'onlineSavingsMsg'
Få mere at vide

MSL2 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16557041
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16557041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16557041 Leverandør Invitrogen Leverandørnr. PA588985

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

MSL2 Polyclonal Antibody

MSL2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen MSL2
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Koncentration 1.24 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MSL2
Genadgangsnr. Q69ZF8, Q9HCI7
Gene Alias E130103E02Rik; E3 ubiquitin-protein ligase MSL2; E3 ubiquitin-protein transferase MSL2; Kiaa1585; male-specific lethal 2 homolog; male-specific lethal 2 homolog (Drosophila); male-specific lethal 2-like 1; Male-specific lethal-2 homolog; Male-specific lethal-2 homolog 1; MSL2; MSL-2; Msl2l1; MSL2-like 1; RGD1310355; ring finger protein 184; Rnf184
Gensymboler MSL2
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 328-577 of human MSL2 (NP_0606032).
Oprensningsmetode Affinity Chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 315959, 55167, 77853
Målarter Human, Mouse, Rat
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.