Få mere at vide
Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform
Artikelnummer. 15287608
Oversigt:
MSIA DART'S, Brugerdefineret
MSIA DART'S, Genopladeligt Stativ
MSIA DART'S, Insulin
MSIA DART'S, Protein A
MSIA DART'S, Protein A/G
MSIA DART'S, Protein G
MSIA DART'S, Streptavidin
Format:
Genopladningsstativ (tips medfølger ikke)
Pakke med 24 spidser
Pakke med 96 spidser
Pakningsstørrelse:
Hver
Pakke med 24
Pakke med 96
Artikelnummer. 15287608
Brand: Thermo Scientific™ 991001096
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Improve hybrid MS bio-analysis with this exclusive workflow solution for proteins and peptides.
Improve hybrid MS bio-analysis with Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette™ and Novus i Platform , an exclusive Thermo Fisher Scientific workflow solution for proteins and peptides. Enabled through proprietary MSIA D.A.R.T.'S™ technologies, these workflows have demonstrated pg/mL level detection while providing simultaneous qualitative/quantitative data content from complex biological matrix, such as human plasma. Ideal for biologic and biomarker bio-analysis, MSIA provides innovative enablement in the areas of PKPD, DMPK, ADME, Biotransformation (DAR and payload determination), as well as routine biomarker analysis.
Next generation hybrid Mass Spectrometry bio-analytical workflows:
  • Enables analysis of proteins of interest down to femtomole level
  • Streamlined workflow for protein qualitative & quantative bio-analysis
  • Employs a proprietary molecular trapping micro-column technology in an easy to use pipette tip housing format
  • Drives antibody/antigen binding kinetics and greatly improves assaying of low-abundance protein from a biological sample
Enabling workflows and technologies for biologic and biomarker research:
  • Immunoaffinity column captures protein of interest with improved signal-to-noise ratio
  • Superior specificity and assaying characteristics compared to conventional approaches
  • Allows for hybrid assaying down to femtomole levels
  • Streptavidin and Protein A, G & A/G MSIA D.A.R.T.'S provide end user flexibility to analyze their specific targets of interest
  • Custom MSIA D.A.R.T.'S are available upon request
  • Optimized workflow solution Insulin (endogenous and therapeutic forms)
Requires:
300 μL pipette such as Thermo Scientific™ Versette™ Liquid Handling Platform, Thermo Scientific™ Finnpipette™ Novus i™ Multichannel pipetters (listed in user manual)
Recommended for:
Hybrid mass spectrometry based assays
For research use only
TRUSTED_SUSTAINABILITY
Tekniske data
Oversigt MSIA DART'S, Insulin
Volumen (metrisk) 300 μL
Format Pakke med 96 spidser
Produktlinje MSIA D.A.R.T.S