missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MS4A3 Antibody (489433) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6247D
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MS4A3 Monoclonal antibody specifically detects MS4A3 in Human samples. It is validated for Flow Cytometry, CyTOF
Tekniske data
| MS4A3 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 932 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 489433 | |
| Flow Cytometry, CyTOF-ready | |
| CD20 antigen homolog, CD20 antigen-like protein, CD20LIgE receptor beta subunit, hematopoietic cell 4 transmembrane protein, Hematopoietic-specific transmembrane protein 4, HTm4, HTM4IgE receptor beta chain, membrane-spanning 4-domains subfamily A member 3, membrane-spanning 4-domains, subfamily A, member 3 (hematopoieticcell-specific) | |
| NS0 mouse myeloma cell line transfected with human MS4A3, Accession # NP_006129 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion