Få mere at vide
Abnova™ MS4A2 293T Cell Transient Overexpression Lysate (Denatured)
Tekniske data
Tekniske data
| Cellelinje | 293T |
| Til brug med (applikation) | Western Blot |
| Oversigt | MS4A2 293T Cell Transient Overexpression Lysate(Denatured) |
| Genadgangsnr. | NM_000139 |
| Molekylvægt (g/mol) | Theoretical M.W. 26.5kDa |
| Plasmid | pCMV-MS4A2 full-length |
| Kvalitetskontrol test | Transient overexpression cell lysate was tested with anti-MS4A2 antibody (Mfr. No. H00002206-D01P) by Western Blot. |
| Mængde | 100 μL |
| Opbevaringsbuffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Værtsarter | Human |
| Vis mere |