Få mere at vide
Tocris Bioscience™ MS 4322
Beskrivelse
MS 4322 is a protein arginine methyltransferase 5 (PRMT5) Degrader (PROTAC™) (DC50 = 1.1 μM, Dmax = 74% in MCF-7 cells). Also inhibits PRMT5 methyltransferase activity (IC50 = 18 nM) and dose-dependently inhibits proliferation of HeLa, A549, A172 and Jurkat cancer cell lines. Comprises a PRMT5 inhibitor linked to an E3 ligase ligand.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-((S)-3-(3,4-Dihydroisoquinolin-2(1H)-yl)-2-hydroxypropyl)-6-((1-((S)-19-((2S,4R)-4-hydroxy-2-(((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)carbamoyl)pyrrolidine-1-carbonyl)-20,20-dimethyl-17-oxo-3,6,9,12,15-pentaoxa-18-azahenicosanoyl)azetidin-3-yl)amin |
| CAS | 2375432-47-4 |
| Mængde | 2 mg |
| Mål | Active Degraders |
| Molekylær formel | C55H76N10O12S |
| Renhed | 98% |