Få mere at vide
Tocris Bioscience™ MRTX 849
Beskrivelse
MRTX 849 is a mutant-selective inhibitor of KRASG12C (IC50 = 142 nM). MRTX 849 covalently binds to KRASG12C at the cysteine at residue 12 and inhibits KRAS-dependent signal transduction by locking the protein in its inactive GDP-bound state. MRTX 849 induces pronounced tumor regression in 17 of 26 (65%) KRASG12C -positive cell lines and patient-derived xenograft models from multiple tumor types. It is orally bioavailable.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-[7-(8-Chloro-1-naphthalenyl)-5,6,7,8-tetrahydro-2-[[(2 S)-1-methyl-2-pyrrolidinyl]methoxy]pyrido[3,4-d]pyrimidin-4-yl]-1-(2-fluoro-1-oxo-2-propen-1-yl)-(2 S)-2-piperazineacetonitrile |
| CAS | 2326521-71-3 |
| Mængde | 50 mg |
| Mål | Ras GTPase Inhibitors |
| Molekylær formel | C32H35ClFN7O2 |
| Renhed | 98% |