missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
MRP1 Antibody (MRP1/1344), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08228IR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MRP1 Monoclonal antibody specifically detects MRP1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
Tekniske data
| MRP1 | |
| Monoclonal | |
| DyLight 755 | |
| ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein | |
| Recombinant full-length human MRP1 protein (Uniprot: P33527) | |
| 100 μg | |
| ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
| 4363 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| MRP1/1344 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion