Få mere at vide
MRL-871, TRC
Artikelnummer. 30475781
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30475781 5 mg -
30469413 1 mg -
30473988 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475781 Leverandør Toronto Research Chemicals Leverandørnr. TRCM7502305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1392809-08-3
Molekylær formel C22H12ClF3N2O3
Molekylvægt (g/mol) 444.79
Synonym 4-[1-[2-Chloro-6-(trifluoromethyl)benzoyl]-1H-indazol-3-yl]benzoic Acid
IUPAC navn 4-(1-(2-chloro-6-(trifluoromethyl)benzoyl)-1H-indazol-3-yl)benzoic acid
SMIL O=C(C1=CC=C(C2=NN(C3=C2C=CC=C3)C(C4=C(C(F)(F)F)C=CC=C4Cl)=O)C=C1)O
Tekniske data
Kemisk navn eller materiale 4-[1-[2-Chloro-6-(trifluoromethyl)benzoyl]-1H-indazol-3-yl]benzoic Acid
Mængde 5 mg
InChI formel InChI=1S/C22H12ClF3N2O3/c23-16-6-3-5-15(22(24,25)26)18(16)20(29)28-17-7-2-1-4-14(17)19(27-28)12-8-10-13(11-9-12)21(30)31/h1-11H,(H,30,31)
Formel vægt 444.05