missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ MRE11A (Human) Recombinant Protein

Product Code. 16137941
missing translation for 'orderingAttributeHoverText'
Mængde:
10 μg
25 μg
This item is not returnable. View return policy

Product Code. 16137941

missing translation for 'mfr': Abnova™ H00004361P01.10ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Human MRE11A full-length ORF ( AAH05241, 1 a.a. - 206 a.a.) recombinant protein with GST-tag at N-terminal.

  • Sequence: MSTADALDDENTFKILVATDIHLGFMEKDAVRGNDTFVTLDEILRLAQENEVDFILLGGDLFHENKPSRKTLHTCLELLRKYCMGDRPVQFEILSDQSVNFGFSKFPWVNYQDGNLNISIPVFSIHGNHDDPTGADALCALDILSCAGFVNHFGRSMSVEKIDISPVLLQKGRTKIALYGLGSIPDERLYRMFVNKKVTMLRPKED

Specifications

Adgangsnummer AAH05241
Gen-id (Entrez) 4361
Navn MRE11 meiotic recombination 11 homolog A (S. cerevisiae)
Fremstillingsmetode Wheat germ expression system
Kvalitetskontrol test 125% SDS-PAGE Stained with Coomassie Blue
Mængde 10 μg
Opbevaringskrav Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias ATLD, HNGS1, MRE11, MRE11B
Gen symbol MRE11A
Arter Wheat Germ (in vitro)
Protein tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.