Få mere at vide
Mrc2 Antibody (Copenhagen Rigshospitalet patent anti-uPARAP) [Janelia Fluor™ 525], Novus Biologicals™
Beskrivelse
Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
Tekniske data
| Antigen | Mrc2 |
| Applikationer | Flow Cytometry, ELISA, Functional Assay |
| Klassifikation | Monoclonal |
| Klon | Copenhagen Rigshospitalet patent anti-uPARAP |
| Konjugeret | Janelia Fluor 525 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein |
| Værtsarter | Mouse |
| Immunogen | MRC2/ CD280 |
| Oprensningsmetode | Protein A purified |
| Vis mere |