Få mere at vide
MPTP hydrochloride, MedChemExpress
Artikelnummer. 16406688
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16406688 10 mg
16412767 50 mg
16426836 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16406688 Leverandør Medchem Express Leverandørnr. HY15608
MedChemExpress MPTP hydrochloride is a brain penetrant dopamine neurotoxin, inducing Parkinson’s Disease. MPTP hydrochloride, a precusor of MPP+, induces apoptosis.
MPTP hydrochloride is a brain penetrant dopamine neurotoxin, inducing Parkinson’s Disease. MPTP hydrochloride, a precusor of MPP+, induces apoptosis.
Tekniske data
Kemisk navn eller materiale MPTP hydrochloride
Fysisk form Solid
CAS 23007-85-4
Farve Off-White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C12H16ClN
Til brug med (applikation) Cancer-programmed cell death
Opløselighedsinformation DMSO : 12 mg/mL (57.22 mM; Need ultrasonic and warming) ∣H2O : 10 mg/mL (47.68 mM; Need ultrasonic)
SMIL CN1CC=C(C2=CC=CC=C2)CC1.[H]Cl
Molekylvægt (g/mol) 209.72
Formel vægt 209.72
Procent renhed 95.0%