Learn More
Invitrogen™ MPIF-1/CCL23 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHCCL23X5
Additional Details : Weight : 0.33820kg
Description
The Human MPIF-1/CCL23 ELISA quantitates Hu MPIF-1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MPIF-1. Principle of the method The Human MPIF-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity on resting T lymphocytes and monocytes, lower activity on neutrophils and no activity on activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. In addition, the product of this gene is a potent agonist of the chemokine (C-C motif) receptor 1. Alternative splicing results in multiple transcript variants that encode different isoforms.Specifications
P55773 | |
7 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
6368 | |
<12% | |
Stop Solution, Biotinylated Detection Antibody, Adhesive Plate Covers, Wash Buffer, SAV-HRP, Chromogen, Assay Diluent Concentrate, Standard, Pre-coated 96 well plates | |
5 x 96 Tests | |
Inflammation | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
8.23 pg/mL to 6000 pg/mL | |
7 pg/mL to 6000 pg/mL | |
Biotin | |
GCPR | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
Human | |
Colorimetric Microplate Reader | |
CK-BETA-8,CKb8,Ckb-8,Ckb-8-1,MIP-3,MIP3,MPIF-1,SCYA23,hmrp-2 A | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
CK-BETA-8, CKb8, Ckb-8, Ckb-8-1, MIP-3, MIP3, MPIF-1, SCYA23, hmrp-2 A | |
1 hr. 20 min. |