missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Endotoxin, Stem Cell Culture

Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation

Brand:  MP Biomedicals™ 0219989690

Additional Details : CAS Number : 9048-46-8 Weight : 0.50000kg

 View more versions of this product

Product Code. 15702272

  • 11560.00 DKK / 500g
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Lipid-rich formulation can be substituted for Albumax™ II.

Physical characteristics:

  • Form: Off white to brown powder
  • Purity:≥98% (w/w)
  • Total Protein: ≥96%
  • pH (4%): 6.5 to 7.5
  • Solubility: Dissolves in 20 minutes at 20° to 25°C
  • Moisture: ≤5% w/w
Microbial evaluation and control:
  • Endotoxin: ≤1EU/mg
  • Mycoplasma: Negative
  • Bioburden: ≤100cfu/g
  • Virus (9CFR): Negative
  • Reported: Lipid content

Certifications: ISO9001, ISO13485, cGMP, EDQM

 

Specifications

Specifications

Mammalian Cell Culture
66
6.5 to 7.5
500g
4°C
Lipid-rich
CellMaxx™ Bovine Albumin, Low Endotoxin
Chromatography
Bovine
Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers