Learn More
MP Biomedicals™ Alcohol Oxidase
From Pichea pastoris E.C.1.1.3.13
Brand: MP Biomedicals™ 0219015505
Additional Details : CAS Number : 9073-63-6 Weight : 0.01000kg
Description
Catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide. Activity: 10-20U per mg protein. Unit Definition: One unit will oxidize 1.0μmole of methanol to formaldehyde per minute at pH 7.5 at 25°C. Solution in sucrose.
Specifications
≥10 U/mg | |
7.2 | |
30% Sucrose, 0.1M Potassium Phosphate Buffer at pH 8 | |
Pichea pastoris | |
Alcohol Oxidase | |
Frozen |
Reddish Brown | |
Colorimetric determination of Blood Alcohol | |
-20°C | |
5 kU | |
≈630000 |