missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ MP A08

Artikelnummer. 18785586
Skift visning
Klik for at se tilgængelige muligheder
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
18785586 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18785586 Leverandør Tocris Bioscience™ Leverandørnr. 5803/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Selective ATP competitive sphingosine kinase 2 and 1 (SphK2/1) inhibitor

MP A08 is a selective ATP competitive sphingosine kinase 2 and 1 (SphK2/1) inhibitor (Ki values are 6.9 and 27 μM, respectively). Exhibits selectivity for SphK 1 and 2 over other kinases. Inhibits cellular S1P production and increases sphingosine and ceramide levels in vitro. Also induces mitochondrial-associated apoptosis. Reduces tumor growth in a human lung adenocarcinoma xenograft mouse model, by inducing apoptosis and suppressing angiogenesis. Cell permeable.
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-Methyl-N-[2-[[[2-[[(4-methylphenyl)sulfonyl]amino]phenyl]imino]methyl]phenyl]benzenesulfonamide
CAS 219832-49-2
Synonym NSC 122314
Mængde 50 mg
Molekylær formel C27H25N3O4S2
Formel vægt 519.64
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.