Få mere at vide
Moxifloxacin Methyl Ester, Mikromol™
Beskrivelse
Moxifloxacin Methyl Ester
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Moxifloxacin Methyl Ester |
| CAS | 721970-35-0 |
| Analyt- eller komponentnavne | Moxifloxacin Methyl Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H26 F N3 O4 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid methyl ester, Moxifloxacin Hydrochloride Impurity H, Moxifloxacin Impurity H |
| InChI formel | InChI=1S/C22H26FN3O4/c1-29-21-18-14(20(27)15(22(28)30-2)10-26(18)13-5-6-13)8-16(23)19(21)25-9-12-4-3-7-24-17(12)11-25/h8,10,12-13,17,24H,3-7,9,11H2,1-2H3/t12-,17+/m0/s1 |
| SMIL | COC(=O)C1=CN(C2CC2)c3c(OC)c(N4C[C@@H]5CCCN[C@@H]5C4)c(F)cc3C1=O |
| IUPAC navn | methyl 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylate |
| Vis mere |