Få mere at vide
Moxifloxacin Hydrochloride, TRC
Artikelnummer. 30464750
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464750 10 mg
1 options
Artikelnummer. 30464750 Leverandør Toronto Research Chemicals Leverandørnr. TRCM74504310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Moxifloxacin hydrochloride
CAS 186826-86-8
Molekylær formel C21 H24 F N3 O4 . Cl H
Mængde 10 mg
Synonym Moxifloxacin hydrochloride, 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid hydrochloride (1:1), (4aS-cis)-1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-(octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl)-4-oxo-3-quinolinecarboxylic acid monohydrochloride, 3-Quinolinecarboxylic acid','1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-','hydrochloride (1:1), 3-Quinolinecarboxylic acid','1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-(octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl)-4-oxo-','monohydrochloride','(4aS-cis)-, 3-Quinolinecarboxylic acid','1-cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-','monohydrochloride (9CI), 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid hydrochloride (1:1), Actira, Avalox, Avelonm, Avelox, BAY 12-8039, Lapinix, Octegra
InChI formel InChI=1S/C21H24FN3O4.ClH/c1-29-20-17-13(19(26)14(21(27)28)9-25(17)12-4-5-12)7-15(22)18(20)24-8-11-3-2-6-23-16(11)10-24;/h7,9,11-12,16,23H,2-6,8,10H2,1H3,(H,27,28);1H/t11-,16+;/m0./s1
SMIL Cl.COc1c(N2C[C@@H]3CCCN[C@@H]3C2)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(=O)O
IUPAC navn 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride
Molekylvægt (g/mol) 437.89
Formel vægt 437.1518