Få mere at vide
Moxifloxacin Hydrochloride, Mikromol™
Beskrivelse
Moxifloxacin Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Moxifloxacin hydrochloride |
| CAS | 186826-86-8 |
| Analyt- eller komponentnavne | Moxifloxacin hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H24 F N3 O4 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Moxifloxacin Hydrochloride, 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid hydrochloride (1:1), (4aS-cis)-1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-(octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl)-4-oxo-3-quinolinecarboxylic acid monohydrochloride, Actira, Avalox, Avelox, BAY 12-8039, Lapinix, Moxifloxacin hydrochloride, Octegra |
| InChI formel | InChI=1S/C21H24FN3O4.ClH/c1-29-20-17-13(19(26)14(21(27)28)9-25(17)12-4-5-12)7-15(22)18(20)24-8-11-3-2-6-23-16(11)10-24;/h7,9,11-12,16,23H,2-6,8,10H2,1H3,(H,27,28);1H/t11-,16+;/m0./s1 |
| SMIL | Cl.COc1c(N2C[C@@H]3CCCN[C@@H]3C2)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(=O)O |
| IUPAC navn | 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride |
| Vis mere |