missing translation for 'onlineSavingsMsg'
Få mere at vide

Moxifloxacin hydrochloride, 97%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J66626.MC

Produktkode 15448059

  • 1850.77 DKK / 100mg

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Moxifloxacin hydrochloride is a broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections. Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.

Solubility
Sparingly soluble in water. Soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

Moxifloxacin hydrochloride
Odorless
100mg
C21H25ClFN3O4
8377447
Sparingly soluble in water; Soluble in DMSO
COC1=C2C(=CC(=C1N3CC4CCCNC4C3)F)C(=O)C(=CN2C5CC5)C(=O)O.Cl
437.896
CHEBI:7008
97%
186826-86-8
Yellow
>110°C (230°F)
MFCD00949117
636°C
IDIIJJHBXUESQI-DFIJPDEKSA-N
7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride
101526
437.89
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.