Få mere at vide
Novus Biologicals™ Mouse XCL1/Lymphotactin ELISA Kit (Chemiluminescence)
Shop alle Bio Techne produkterTekniske data
Tekniske data
| Analyseområde | 15.63 to 1000 pg/mL |
| Assay følsomhed | 9.38 pg/mL |
| Produkttype | ELISA Kit (Chemiluminescence) |
| Prøvetype | Serum, Plasma, Biological Fluids |
| Specificitet | This kit recognizes Mouse Lptn/XCL1 in samples. No significant cross-reactivity or interference between Mouse Lptn/XCL1 and analogues was observed. |
| Til brug med (applikation) | ELISA |
| Gen-id (Entrez) | 6375 |
| Gen symbol | XCL1 |
| Mængde | 1 sæt |
| Forskningsdisciplin | Biologically Active Proteins, Immunology, Innate Immunity |
| Vis mere |