Få mere at vide
Novus Biologicals™ Mouse XCL1/Lymphotactin ELISA Kit (Chemiluminescence)
Artikelnummer. 18643840 Shop alle Bio Techne produkter
Skift visning
Mængde:
1 sæt
Artikelnummer. Mængde
18643840 1 sæt
1 options
Artikelnummer. 18643840 Leverandør Novus Biologicals™ Leverandørnr. NBP268055
Kit (Chemiluminescence)
TRUSTED_SUSTAINABILITY
Tekniske data
Analyseområde 15.63 to 1000 pg/mL
Assay følsomhed 9.38 pg/mL
Produkttype ELISA Kit (Chemiluminescence)
Prøvetype Serum, Plasma, Biological Fluids
Specificitet This kit recognizes Mouse Lptn/XCL1 in samples. No significant cross-reactivity or interference between Mouse Lptn/XCL1 and analogues was observed.
Til brug med (applikation) ELISA
Gen-id (Entrez) 6375
Gen symbol XCL1
Mængde 1 sæt
Forskningsdisciplin Biologically Active Proteins, Immunology, Innate Immunity
Opbevaringskrav Storage of components varies. See protocol for specific instructions.
Synonym ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 A, SCM1A, SCYC1SCM1, single cysteine motif 1 A, small inducible cytokine subfamily C, member 1 (lymphotactin)
Mål Mouse