Få mere at vide
R&D Systems™ Mouse TSLP (NP_067342.1) VersaClone cDNA
Tekniske data
Tekniske data
| Gene | TSLP |
| Adgangsnummer | NP_067342.1 |
| Gen-id (Entrez) | 85480 |
| Gene Alias | thymic stromal lymphopoietin |
| Indhold og opbevaring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Mængde | 10 μg |