missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Mouse TNF-alpha Quantikine HS ELISA Kit
Klik for at se tilgængelige muligheder
Mængde:
1 sæt
Beskrivelse
Indeholder
E. coli-expressed recombinant mouse TNF-alpha and antibodies raised against the recombinant factor
The Mouse TNF-alpha Quantikine HS ELISA Kit quantifies mouse TNF-alpha in serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Tekniske data
Tekniske data
| Oversigt | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, tnfa, TNF-a, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Produkttype | ELISA Kit |
| Mængde | 1 sæt |
| Arter | Mouse |
| Assay følsomhed | 0.295 pg/mL |
| Analyseområde | 0.78 to 50 pg/mL |
| Krydsreaktivitet | <0.5% cross-reactivity observed with available related molecules, <50% cross-species reactivity observed with species tested |
| Til brug med (applikation) | ELISA |
| Gen-id (Entrez) | 7124 |
| Prøvetype | Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL) |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion