Få mere at vide
R&D Systems™ Mouse TL1A/TNFSF15 DuoSet ELISA
Artikelnummer. 30021868
Skift visning
Format:
96-brønds plader
Artikelnummer. Format
30021868 96-brønds plader
1 options
Artikelnummer. 30021868 Leverandør R&D Systems™ Leverandørnr. DY189605
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
For the determination of natural and recombinant TL1A in mouse. R&D Systems Mouse TL1A/TNFSF15 DuoSet ELISA Kit is a solid phase sandwich ELISA suitable for the analysis of cell culture supernate, serum and plasma samples.
The Mouse TL1A/TNFSF15 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse TL1A/TNFSF15.
TRUSTED_SUSTAINABILITY
Tekniske data
Produkttype ELISA Kit
Arter Mouse
Bindende mål TL1A
Format 96-brønds plader
Til brug med (applikation) ELISA
Gen-id (Entrez) 9966
Prøvetype Cell Culture Supernatant, Serum, Plasma
Indhold og opbevaring Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Research Use Only.