missing translation for 'onlineSavingsMsg'
Få mere at vide

Mouse TGF-beta RII APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. 18718148
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18718148 25 Tests
18748817 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18718148 Leverandør R&D Systems Leverandørnr. FAB532A025

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Goat Polyclonal Antibody has been used in 5 publications

TGF-beta RII Polyclonal specifically detects TGF-beta RII in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen TGF-beta RII
Applikationer Flow Cytometry
Klassifikation Polyclonal
Konjugeret APC
Fortynding Flow Cytometry 10 uL/10^6 cells
Formulering Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Genadgangsnr. Q62312
Gene Alias AAT3, EC 2.7.11, EC 2.7.11.30, FAA3, HNPCC6, LDS1B, LDS2B, MFS2, RIIC, TAAD2, tbetaR-II, TGF-beta receptor type II, TGF-beta receptor type IIB, TGF-beta receptor type-2, TGF-beta type II receptor, TGFbetaRII, TGFbeta-RII, TGFBR2, TGF-bRII, TGFR-2, transforming growth factor beta receptor type IIC, transforming growth factor, beta receptor II (70/80kDa), transforming growth factor, beta receptor II (70/80kDa) isoform 1, transforming growth factor, beta receptor II (70/80kDa) isoform 2, transforming growth factor, beta receptor II (70-80kD), Transforming growth factor-beta receptor type II
Gensymboler TGFBR2
Værtsarter Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TGF-beta RII and S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RII Ile24-Asp184 Accession # Q62312
Oprensningsmetode Affinity Purified
Mængde 25 Tests
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 7048
Testspecificitet Detects mouse TGF-beta RII in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) TGF-beta RII is observed and less than 1% cross-reactivity with recombinant mouse TGF-beta RI and rhTGF-beta RIII is observed.
Målarter Mouse
Indhold og opbevaring Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.