missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Mouse SLITRK1 (NP_951020) VersaClone cDNA

Artikelnummer. 18713798
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18713798 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18713798 Leverandør R&D Systems™ Leverandørnr. RDC1637
  • ONLINE EXCLUSIVE PRICE
    5975.00 DKK
    5649.00 DKK / 10µg
    Spare 326.00 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 14-10-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Mouse SLITRK1 (NP_951020) VersaClone cDNA clone contains the complete ORF for SLITRK1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLITRK1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Ordreinfo

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Tekniske data

Gene SLITRK1
Adgangsnummer NP_951020
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10mM Tris-Cl, pH 8.308
Indsæt type 2104bp
Gen-id (Entrez) 114798
Gene Alias FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Mouse
Mængde 10 μg

For Reasearch Use Only. Not for use in Humans.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.