missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Mouse SCF, Animal-Free Recombinant Protein, PeproTech®

Artikelnummer. 17883783
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17883783 1 mg
17883033 100 μg
17853133 10 μg
17853263 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Vare(r) i denne gruppe er en alternativ mulighed til delnummer< delnummer>
Artikelnummer. 17883783 Leverandør Gibco™ Leverandørnr. AF250031MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Vare(r) i denne gruppe er en alternativ mulighed til delnummer< delnummer>

Recombinant Protein

Recombinant Murine SCF is an 18.3 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P20826
Til brug med (applikation) Functional Assay
Formulering protein with no preservative
Gen-id (Entrez) 17311
Molekylvægt (g/mol) 18.3 kDa
Navn Mouse SCF, Animal-Free
Mængde 1 mg
Kilde E. coli
Regulatorisk status RUO
Endotoksin koncentration <0.1 EU/ μg
Gene Alias 710-712; blz; c-Kit ligand; Clo; cloud gray; Con; contrasted; CSF; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; Gb; grizzle-belly; hematopoietic growth factor KL; H-SCF; Kit ligand; Kitl; Kitlg; kit-ligand; KL-1; Mast cell growth factor; MGF; M-SCF; Processed kit ligand; SCF; SF; SHEP7; sKITLG; Sl; SLF; Soluble KIT ligand; Steel factor; steel factor/kit ligand; stem cell factor; stem cell factor KL-1; stem cell factor, CSF
Fælles navn SCF
Gen symbol Kitl
Biologisk aktivitet The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 10.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^5 units/mg
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA
Indhold og opbevaring -20°C
Udtrykssystem E. coli
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 98% by SDS-PAGE gel and HPLC analyses.
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.