missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Mouse/Rat IL-17F Quantikine ELISA Kit

Product Code. 30001846
Click to view available options
Mængde:
1 sæt
This item is not returnable. View return policy

Product Code. 30001846

Brand: R&D Systems™ M17F0

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Mouse/Rat IL-17F Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat IL-17F Quantikine ELISA Kit quantifies mouse, rat IL-17F in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Mængde 1 sæt
Arter Mouse, Rat
Konjugeret HRP
Bindende mål IL-17F
Assay følsomhed 1.74 pg/mL
Analyseområde 15.60 to 1000 pg/mL
Til brug med (applikation) ELISA
Gen-id (Entrez) 112744
Prøvetype Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL)
Test tid 4.5 hr.
Synonym Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F
Indhold og opbevaring Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.