Få mere at vide
R&D Systems™ Mouse Proprotein Convertase 9 DuoSet ELISA
Beskrivelse
The Mouse Proprotein Convertase 9 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Proprotein Convertase 9/PCSK9.
Tekniske data
Tekniske data
| Mængde | 1 sæt (til 15 tallerkener) |
| Arter | Mouse |
| Konjugeret | Biotin |
| Analyseområde | 31.20 to 2000 pg/mL |
| Til brug med (applikation) | ELISA |
| Gen-id (Entrez) | 100102 |
| Synonym | EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1 convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 |
| Indhold og opbevaring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |