Få mere at vide
Mouse Podocalyxin Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 32 publications
Brand: R&D Systems AF1556
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Podocalyxin Like Polyclonal specifically detects Podocalyxin Like in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Tekniske data
| Podocalyxin Like | |
| Polyclonal | |
| Unconjugated | |
| Q9R0M4 | |
| PODXL | |
| Mouse myeloma cell line NS0-derived recombinant mouse Podocalyxin Ser21-Arg402 Accession # Q9R0M4 | |
| 100 μg | |
| Primary | |
| Detects mouse Podocalyxin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Podocalyxin and rhEndoglycan is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
| GCTM, Gp200MGC138240, PCLP, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, POD XL, podocalyxin-like, Podocalyxin-like protein 1, PODXL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5420 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |