Få mere at vide
R&D Systems™ Mouse OLT-2/TARM1 (NP_796337) VersaClone cDNA
Artikelnummer. 18760939
Skift visning
Mængde:
10 μg
Artikelnummer. Mængde
18760939 10 μg
1 options
Artikelnummer. 18760939 Leverandør R&D Systems™ Leverandørnr. RDC2318
  • ONLINE EXCLUSIVE PRICE
    2605.00 DKK
    2460.15 DKK / 10µg
    Spare 144.85 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 28-08-2026
Læg i indkøbskurv
Læg i indkøbskurv
R&D Systems™ Mouse OLT-2/TARM1 (NP_796337) VersaClone cDNA may act as receptor.
The Mouse OLT-2/TARM1 (NP_796337) VersaClone cDNA clone contains the complete ORF for OLT-2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The OLT-2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY
Tekniske data
Gene mTarm1
Adgangsnummer NP_796337
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl
Indsæt type 880bp
pH 8.5
Gen-id (Entrez) 441864
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Mouse
Mængde 10 μg