Learn More
Gibco™ Mouse M-CSF Recombinant Protein, PeproTech®
Recombinant Protein
642.00 DKK - 34260.00 DKK
Specifications
Accession Number | P07141 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 12977 |
Molecular Weight (g/mol) | 36.4 kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17812423
|
Gibco™
315-02-100UG |
100 ÎĽg |
6830.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17872233
|
Gibco™
315-02-10UG |
10 ÎĽg |
1556.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17812133
|
Gibco™
315-02-1MG |
1 mg |
34260.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17812113
|
Gibco™
315-02-250UG |
250 ÎĽg |
12010.00 DKK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17822333
|
Gibco™
315-02-2UG |
2 ÎĽg |
642.00 DKK
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17862003
|
Gibco™
315-02-500UG |
500 ÎĽg |
20530.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17882003
|
Gibco™
315-02-50UG |
50 ÎĽg |
4115.00 DKK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant Murine M-CSF is a 36.4 kDa homodimeric protein consisting of two 156 amino acid polypeptide subunits. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.Specifications
P07141 | |
protein with no preservative | |
36.4 kDa | |
E. coli | |
<1 EU/ ÎĽg | |
M-CSF | |
The ED50 as determined by the dose-dependent stimulation of the proliferation of M-NFS-60 cells is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg | |
Recombinant | |
-20°C | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
12977 | |
Mouse M-CSF | |
RUO | |
C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1 | |
Csf1 | |
Unconjugated | |
MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP | |
E. coli | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.